Bis(2,2'-bipyridine-κN1,κN1')(dipyrido[3,2-a:2',3'-c]phenazine-κN4,κN5)ruthenium(2+) 98%

Supplier: Ambeed
Danger

A728925-100MG A728925-1G A728925-250MG
77267-736EA 633 USD
77267-736 77267-738 77267-740
Bis(2,2'-bipyridine-κN1,κN1')(dipyrido[3,2-a:2',3'-c]phenazine-κN4,κN5)ruthenium(2+) 98%
Bis(2,2'-bipyridine-κN1,κN1')(dipyrido[3,2-a:2',3'-c]phenazine-κN4,κN5)ruthenium(2+)

Precaución: for Research Use Only

Formula: C₃₈H₂₆N₈Ru
MW: 695.73 g/mol
CAS Number: 87564-74-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR