3-Methylenecyclobutanecarboxylic acid 98%

Supplier: Ambeed
Danger

A727975-1G A727975-5G
77281-016EA 38 USD
77281-016 77552-252
3-Methylenecyclobutanecarboxylic acid 98%
3-Methylenecyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C6H8O2
MDL Number: MFCD00101901
CAS Number: 15760-36-8
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR