4-(Trifluoromethyl)-1H-pyrazole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A727922-1G A727922-250MG
77243-748EA 795.16 USD
77243-748 77243-750
4-(Trifluoromethyl)-1H-pyrazole-3-carboxylic acid 96%
4-(Trifluoromethyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
MDL Number: MFCD22030979
CAS Number: 716362-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR