(But-3-yn-1-yloxy)(tert-butyl)dimethylsilane 98%

Supplier: Ambeed
Danger

Synonyms: 4-(tert-Butyldimethylsilyloxy)-1-butyne, (But-3-yn-1-yloxy)(tert-butyl)dimethylsilane

A727806-1G A727806-25G A727806-5G
77293-428EA 20.6 USD
77293-428 77293-430 77293-432
(But-3-yn-1-yloxy)(tert-butyl)dimethylsilane 98%
(But-3-yn-1-yloxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₂₀OSi
MW: 184.36 g/mol
MDL Number: MFCD03427046
CAS Number: 78592-82-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR