2,4-Dimethylbenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2,4-dimethylbenzene

A727515-10G A727515-1G A727515-250MG A727515-25G A727515-5G
77255-424EA 285.31 USD
77255-424 77255-426 77255-428 77255-430 77255-432
2,4-Dimethylbenzyl bromide 97%
2,4-Dimethylbenzyl bromide

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.09 g/mol
MDL Number: MFCD03844740
CAS Number: 78831-87-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR