4-Methoxy-3-(trifluoromethyl)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A727381-10G
A727381-1G
A727381-250MG
A727381-25G
A727381-5G
77267-726EA
63
USD
77267-726
77267-728
77267-730
77267-732
77267-734
4-Methoxy-3-(trifluoromethyl)benzaldehyde 98%
4-Methoxy-3-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₇F₃O₂ |
MDL Number:
MFCD01091012 CAS Number: 50823-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...