(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-1-[(S)-2-(Dicyclohexylphosphino)-ferrocenyl]ethyldicyclohexylphosphine

A727254-100MG A727254-1G A727254-250MG A727254-5G
77255-416EA 47.38 USD
77255-416 77255-418 77255-420 77255-422
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥98%, ee 99%
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine

Precaución: for Research Use Only

Formula: C₃₆H₅₆FeP₂
MW: 606.6400 g/mol
MDL Number: MFCD00800279
CAS Number: 246231-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR