2',4'-Dihydroxychalcone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Dihydroxyphenyl)-3-phenyl-2-propenone, 1-(2,4-Dihydroxyphenyl)-3-phenylprop-2-en-1-one, 2',4'-Dihydroxybenzylidenacetophenone

A727107-100MG A727107-1G A727107-250MG
77255-402EA 163.77 USD
77255-402 77255-404 77255-406
2',4'-Dihydroxychalcone 95%
2',4'-Dihydroxychalcone

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₃
MW: 240.26 g/mol
MDL Number: MFCD00180050
CAS Number: 1776-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR