2-Bromo-6-chloro-3-iodo-5-methylpyridine 95%

Supplier: Ambeed
Warning

A726956-1G A726956-250MG A726956-5G
77267-720EA 475.86 USD
77267-720 77267-722 77267-724
2-Bromo-6-chloro-3-iodo-5-methylpyridine 95%
2-Bromo-6-chloro-3-iodo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClIN
MW: 332.36 g/mol
MDL Number: MFCD22572506
CAS Number: 1822782-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR