2-Bromo-3',5'-bis(trifluoromethyl)acetophenone 95%

Supplier: Ambeed
Danger

Synonyms: 1-(3,5-Bis(trifluoromethyl)phenyl)-2-bromoethanone, 3,5-Bis(trifluoromethyl)phenacyl bromide

A726446-1G A726446-250MG A726446-5G
77281-006EA 38.11 USD
77281-006 77281-008 77281-010
2-Bromo-3',5'-bis(trifluoromethyl)acetophenone 95%
2-Bromo-3',5'-bis(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₆O
MW: 335.0405 g/mol
MDL Number: MFCD00792434
CAS Number: 131805-94-2
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR