(2-Chloro-6-methoxypyridin-4-yl)methanamine 97%

Supplier: Ambeed
Danger

A725635-100MG A725635-1G A725635-250MG
77243-730EA 67 USD
77243-730 77243-732 77243-734
(2-Chloro-6-methoxypyridin-4-yl)methanamine 97%
(2-Chloro-6-methoxypyridin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MDL Number: MFCD18204705
CAS Number: 1256788-06-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR