Methyl (2E)-4-methylpent-2-enoate 96%

Supplier: Ambeed
Danger

Synonyms: Methyl (E)-3-isopropylacrylate, Methyl trans-4-methylpent-2-enoate, (E)-Methyl 4-methylpent-2-enoate

A724992-1G A724992-5G
77267-718EA 9.27 USD
77267-718 77617-970
Methyl (2E)-4-methylpent-2-enoate 96%
Methyl (2E)-4-methylpent-2-enoate

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.169 g/mol
MDL Number: MFCD00137612
CAS Number: 20515-15-5
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR