5-Chloro-2-hydroxybenzene-1,3-dicarbaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-2-hydroxyisophthalaldehyde
A724965-100MG
A724965-1G
A724965-250MG
77255-396EA
76.22
USD
77255-396
77255-398
77255-400
5-Chloro-2-hydroxybenzene-1,3-dicarbaldehyde 95%
5-Chloro-2-hydroxybenzene-1,3-dicarbaldehyde
Precaución: for Research Use Only
Formula:
C₈H₅ClO₃ MW: 184.577 g/mol |
MDL Number:
MFCD00067051 CAS Number: 32596-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...