(11bR)-4-Hydroxy-2,6-bis(2,4,6-triisopropylphenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥98%, ee 99%

Supplier: Ambeed
Warning

A724793-1G A724793-250MG
77255-394EA 414.99 USD
77255-394 77617-968
(11bR)-4-Hydroxy-2,6-bis(2,4,6-triisopropylphenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥98%, ee 99%
(11bR)-4-Hydroxy-2,6-bis(2,4,6-triisopropylphenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₅₀H₆₅O₄P
MDL Number: MFCD30541785
CAS Number: 929294-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR