APY29 98%

Supplier: Ambeed
Warning

Synonyms: N2-(1H-Benzo[d]imidazol-6-yl)-N4-(5-cyclopropyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine

A724754-10MG A724754-1MG A724754-50MG A724754-5MG
77243-724EA 60.77 USD
77243-724 77243-726 77243-728 77617-966
APY29 98%
APY29

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₈
MW: 332.36 g/mol
MDL Number: MFCD28023582
CAS Number: 1216665-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR