12H-Benzofuro[2,3-a]carbazole 97%

Supplier: Ambeed
Warning

A724699-1G A724699-5G
77281-000EA 50.47 USD
77281-000 77281-002
12H-Benzofuro[2,3-a]carbazole 97%
12H-Benzofuro[2,3-a]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁NO
MW: 257.28 g/mol
MDL Number: MFCD32670210
CAS Number: 1338919-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR