1-Ethyl-3-methylimidazoliumtetrafluoroborate 97%

Supplier: Ambeed
Danger

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

A724626-5G A724626-25G
77243-720EA 11.33 USD
77243-720 77243-722 77308-208
1-Ethyl-3-methylimidazoliumtetrafluoroborate 97%
1-Ethyl-3-methylimidazoliumtetrafluoroborate

Precaución: for Research Use Only

Formula: C₆H₁₁BF₄N₂
MDL Number: MFCD00216668
CAS Number: 143314-16-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR