5-(2-Chloropyridin-4-yl)-3-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole 97%

Supplier: Ambeed
Warning

A724189-100MG A724189-1G A724189-250MG
77243-714EA 99.91 USD
77243-714 77243-716 77243-718
5-(2-Chloropyridin-4-yl)-3-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole 97%
5-(2-Chloropyridin-4-yl)-3-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole

Precaución: for Research Use Only

Formula: C₁₄H₇ClF₃N₃O
MW: 325.67 g/mol
MDL Number: MFCD00833044
CAS Number: 266692-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR