2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%

Supplier: Ambeed
Warning

A724153-1G A724153-25G A724153-5G
77255-388EA 15 USD
77255-388 77255-390 77255-392
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

Precaución: for Research Use Only

Formula: C₆H₆F₈O₂
MDL Number: MFCD00069087
CAS Number: 355-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR