3'-Bromoacetanilide 98%

Supplier: Ambeed
Warning

A724064-100G A724064-1G
77280-996EA 202 USD
77280-996 77280-998
3'-Bromoacetanilide 98%
3'-Bromoacetanilide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD00461391
CAS Number: 621-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR