LJI-308 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-4-(4-(4-morpholinophenyl)pyridin-3-yl)phenol, 2,6-Difluoro-4-[4-[4-(4-morpholinyl)phenyl]-3-pyridinyl]-phenol

A723919-10MG A723919-1MG A723919-2MG A723919-5MG
77243-706EA 391.4 USD
77243-706 77243-708 77243-710 77243-712
LJI-308 98%
LJI-308

Precaución: for Research Use Only

Formula: C₂₁H₁₈F₂N₂O₂
MW: 368.38 g/mol
MDL Number: MFCD29905461
CAS Number: 1627709-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR