2,4,6-Trimethylphenol 98%

Supplier: Ambeed
Danger

A723689-10G A723689-25G A723689-100G
77255-384EA 14 USD
77255-384 77255-386 77747-431
2,4,6-Trimethylphenol 98%
2,4,6-Trimethylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00002235
CAS Number: 527-60-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR