2-Amino-3-fluoroisonicotinic acid 97%

Supplier: Ambeed
Warning

A723501-100MG A723501-250MG
77243-698EA 20 USD
77243-698 77243-700
2-Amino-3-fluoroisonicotinic acid 97%
2-Amino-3-fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MDL Number: MFCD18257286
CAS Number: 1256809-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR