Ethylene Glycol bis(2-aminophenyl) ether 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(2-aminophenoxy)ethane, 2,2'-[Ethane-1,2-diylbis(oxy)]dianiline, 1,2-Bis(o-aminophenoxy)ethane

A723467-10G A723467-5G
77255-380EA 19 USD
77255-380 77255-382
Ethylene Glycol bis(2-aminophenyl) ether 97%
Ethylene Glycol bis(2-aminophenyl) ether

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
MDL Number: MFCD00087648
CAS Number: 52411-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR