3-Methylquinoxalin-2-amine 95%

Supplier: Ambeed
Warning

A723358-1G A723358-250MG
77255-370EA 294.58 USD
77255-370 77255-372
3-Methylquinoxalin-2-amine 95%
3-Methylquinoxalin-2-amine

Precaución: for Research Use Only

Formula: C₉H₉N₃
MW: 159.18 g/mol
MDL Number: MFCD00234542
CAS Number: 34972-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR