1-Bromo-2,2-dimethoxypropane 97%

Supplier: Ambeed
Danger

A723126-1G A723126-25G A723126-100G A723126-5G
77243-696EA 11.33 USD
77243-696 77552-236 77747-429 77747-430
1-Bromo-2,2-dimethoxypropane 97%
1-Bromo-2,2-dimethoxypropane

Precaución: for Research Use Only

Formula: BrH₂CC(OCH₃)₂CH₃
MW: 183.05 g/mol
MDL Number: MFCD00000208
CAS Number: 126-38-5
UN: 3271
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR