1-(2,4,6-Trimethylphenyl)-3-(adamantyl)imidazolium chloride 98%

Supplier: Ambeed
Warning

A723011-100MG A723011-1G A723011-250MG A723011-5G
77255-356EA 63.86 USD
77255-356 77255-358 77255-360 77255-362
1-(2,4,6-Trimethylphenyl)-3-(adamantyl)imidazolium chloride 98%
1-(2,4,6-Trimethylphenyl)-3-(adamantyl)imidazolium chloride

Precaución: for Research Use Only

Formula: C₂₂H₂₉ClN₂
MW: 356.93 g/mol
MDL Number: MFCD28139678
CAS Number: 1583244-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR