4-(2-((6-Ethoxy-1H-benzo[d]imidazol-2-yl)thio)ethyl)morpholine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fabomotizole HCl
A722960-10MG
A722960-1MG
A722960-25MG
A722960-50MG
A722960-5MG
77243-686EA
32.96
USD
77243-686
77243-688
77243-690
77243-692
77243-694
4-(2-((6-Ethoxy-1H-benzo[d]imidazol-2-yl)thio)ethyl)morpholine hydrochloride 98%
4-(2-((6-Ethoxy-1H-benzo[d]imidazol-2-yl)thio)ethyl)morpholine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₂₂ClN₃O₂S MW: 343.87 g/mol |
MDL Number:
MFCD28167801 CAS Number: 173352-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...