8-Chloro-N-(4-(trifluoromethoxy)phenyl)quinolin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: Obefazimod

A722436-10MG A722436-1MG A722436-5MG A722436-50MG
77267-712EA 42.23 USD
77267-712 77267-714 77267-716 77749-471
8-Chloro-N-(4-(trifluoromethoxy)phenyl)quinolin-2-amine 98%
8-Chloro-N-(4-(trifluoromethoxy)phenyl)quinolin-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₀ClF₃N₂O
MW: 338.71 g/mol
MDL Number: MFCD31657325
CAS Number: 1258453-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR