1-Phenyl-1,3-butadiene 95%

Supplier: Ambeed
Warning

A722186-1G A722186-250MG A722186-5G
77243-680EA 147.29 USD
77243-680 77243-682 77243-684
1-Phenyl-1,3-butadiene 95%
1-Phenyl-1,3-butadiene

Precaución: for Research Use Only

Formula: C₁₀H₁₀
MW: 130.18 g/mol
CAS Number: 1515-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR