3-Iodo-7-nitroindazole 95%

Supplier: Ambeed
Warning

Synonyms: 3-Iodo-7-nitro-1H-indazole

A722059-10G A722059-1G
77280-980EA 294.58 USD
77280-980 77280-982
3-Iodo-7-nitroindazole 95%
3-Iodo-7-nitroindazole

Precaución: for Research Use Only

Formula: C₇H₄IN₃O₂
MW: 289.03 g/mol
MDL Number: MFCD09027004
CAS Number: 864724-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR