5-Bromo-2-cyanonicotinic acid 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-3-carboxypicolinonitrile, 5-Bromo-2-cyanopyridine-3-carboxylic acid, 5-Bromo-3-carboxy-2-cyanopyridine

A721695-100MG A721695-250MG A721695-50MG
77243-670EA 209 USD
77243-670 77243-672 77243-674
5-Bromo-2-cyanonicotinic acid 97%
5-Bromo-2-cyanonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
MW: 227.015 g/mol
MDL Number: MFCD08543463
CAS Number: 914637-97-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR