(S)-3,3'-Bis[4-(1-naphthalenyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate 98%

Supplier: Ambeed
Warning

A721446-100MG A721446-250MG
77255-348EA 114.33 USD
77255-348 77255-350
(S)-3,3'-Bis[4-(1-naphthalenyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate 98%
(S)-3,3'-Bis[4-(1-naphthalenyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate

Precaución: for Research Use Only

Formula: C₅₂H₃₃O₄P
MW: 752.78 g/mol
CAS Number: 871130-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR