Potassium O,O-diethyl phosphorothioate 98%

Supplier: Ambeed
Warning

A721290-1G A721290-25G A721290-5G
77255-342EA 24.72 USD
77255-342 77255-344 77255-346
Potassium O,O-diethyl phosphorothioate 98%
Potassium O,O-diethyl phosphorothioate

Precaución: for Research Use Only

Formula: C₄H₁₀KO₃PS
MW: 208.25 g/mol
MDL Number: MFCD00674011
CAS Number: 5871-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR