5-Bromo-2-furoic acid hydrazide 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromofuran-2-carbohydrazide

A721131-1G A721131-5G
77293-410EA 19 USD
77293-410 77293-412
5-Bromo-2-furoic acid hydrazide 95%
5-Bromo-2-furoic acid hydrazide

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MDL Number: MFCD01310787
CAS Number: 89282-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR