4'-Nitro-3'-(trifluoromethyl)acetanilide 97%

Supplier: Ambeed
Danger

Synonyms: N-(4-Nitro-3-(trifluoromethyl)phenyl)acetamide

A720567-10G A720567-1G A720567-25G A720567-5G
77267-704EA 70.04 USD
77267-704 77267-706 77267-708 77267-710
4'-Nitro-3'-(trifluoromethyl)acetanilide 97%
4'-Nitro-3'-(trifluoromethyl)acetanilide

Precaución: for Research Use Only

Formula: C₉H₇F₃N₂O₃
MW: 248.1600 g/mol
MDL Number: MFCD00017994
CAS Number: 393-12-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR