2-Chloro-4,6-di(naphthalen-2-yl)-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A720503-1G A720503-250MG A720503-5G
77255-330EA 18.01 USD
77255-330 77255-332 77255-334
2-Chloro-4,6-di(naphthalen-2-yl)-1,3,5-triazine 97%
2-Chloro-4,6-di(naphthalen-2-yl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₃H₁₄ClN₃
MDL Number: MFCD28716938
CAS Number: 1247124-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR