4,5-Dibromo-2-fluorotoluene 95%

Supplier: Ambeed
Warning

A720260-10G A720260-1G A720260-25G A720260-5G
77293-396EA 37 USD
77293-396 77293-398 77293-400 77293-402
4,5-Dibromo-2-fluorotoluene 95%
4,5-Dibromo-2-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MDL Number: MFCD00143234
CAS Number: 202982-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR