3-(2,4-Dimethoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,4-Dimethoxyphenyl)propanoic acid

A719366-10G A719366-1G A719366-5G
77280-954EA 369.77 USD
77280-954 77280-956 77280-958
3-(2,4-Dimethoxyphenyl)propionic acid 98%
3-(2,4-Dimethoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
MDL Number: MFCD00598205
CAS Number: 22174-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR