1-Butyl-pyrrole-2,5-dione 95+%

Supplier: Ambeed
Danger

A719354-1G A719354-100MG
77243-640EA 195 USD
77243-640 77747-424
1-Butyl-pyrrole-2,5-dione 95+%
1-Butyl-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MDL Number: MFCD00042756
CAS Number: 2973-09-3
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR