2-Bromo-7-nitroquinoline 95%

Supplier: Ambeed
Warning

A719010-1G A719010-250MG
77267-698EA 350.2 USD
77267-698 77267-700
2-Bromo-7-nitroquinoline 95%
2-Bromo-7-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O₂
MW: 253.05 g/mol
MDL Number: MFCD28048439
CAS Number: 1822819-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR