4'-Chloro-3'-(trifluoromethyl)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-(trifluoromethyl)acetophenone

A718925-100G A718925-25G
77267-694EA 162.74 USD
77267-694 77267-696
4'-Chloro-3'-(trifluoromethyl)acetophenone 98%
4'-Chloro-3'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
MDL Number: MFCD03094146
CAS Number: 129825-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR