3-Aminobenzene-1-sulfonyl fluoride 97%

Supplier: Ambeed
Danger

A718732-100MG A718732-250MG
77255-306EA 52 USD
77255-306 77552-212
3-Aminobenzene-1-sulfonyl fluoride 97%
3-Aminobenzene-1-sulfonyl fluoride

Precaución: for Research Use Only

Formula: C₆H₆FNO₂S
MDL Number: MFCD00047847
CAS Number: 368-50-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR