UNC-3866 99+%

Supplier: Ambeed
Warning

Synonyms: N-[4-(1,1-dimethylethyl)benzoyl]-L-phenylalanyl-L-alanyl-L-leucyl-N6,N6-diethyl-L-lysyl-L-serine, UNC3866, Methyl ester, (3S,6S,9S,12S,15S)-Methyl 3-benzyl-1-(4-(tert-butyl)phenyl)-12-(4-(diethylamino)butyl)-15-(hydroxymethyl)-9-isobutyl-6-methyl-1,4,7,10,13-pentaoxo-2,5,8,11,14-pentaazahexadecan-16-oate

A718589-10MG A718589-1MG A718589-25MG A718589-50MG A718589-5MG
77267-684EA 43 USD
77267-684 77267-686 77267-688 77267-690 77267-692
UNC-3866 99+%
UNC-3866

Precaución: for Research Use Only

Formula: C₄₃H₆₆N₆O₈
MW: 795.04 g/mol
MDL Number: MFCD30536205
CAS Number: 1872382-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR