3,5-Dimethylisoxazole-4-carbonyl chloride 98%

Supplier: Ambeed
Danger

A718537-1G A718537-25G A718537-5G
77280-942EA 14.42 USD
77280-942 77280-944 77280-946
3,5-Dimethylisoxazole-4-carbonyl chloride 98%
3,5-Dimethylisoxazole-4-carbonyl chloride

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂
MW: 159.57 g/mol
MDL Number: MFCD00052555
CAS Number: 31301-45-8
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR