5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] 95% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: NHS-Rhodamine, NHS-Rhoda, TAMRA SE, 5 and 6 isomers, TMR-SE, 5(6)-Carboxytetramethylrhodamine N-succinimidyl ester, 5(6)-TAMRA N-succinimidyl ester

A718529-100MG A718529-1MG A718529-50MG A718529-5MG
77267-678EA 221 USD
77267-678 77267-680 77267-682 77749-467
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] 95% (mixture of isomers)
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

Precaución: for Research Use Only

Formula: C₂₉H₂₅N₃O₇
MDL Number: MFCD00077324
CAS Number: 150408-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR