Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-1-enecarboxylate 97%
Supplier: Ambeed
|
Warning
|
A718494-1G
A718494-250MG
77243-624EA
329.6
USD
77243-624
77243-626
Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-1-enecarboxylate 97%
Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-1-enecarboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₂₅BO₄ MW: 280.16 g/mol |
MDL Number:
MFCD11520539 CAS Number: 497959-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...