Allyl phenyl carbonate 97%

Supplier: Ambeed
Warning

Synonyms: Allyl Phenyl Carbonate, ['Carbonic acid Allyl Phenyl Ester']

A718187-1G A718187-25G A718187-5G
77267-672EA 18.54 USD
77267-672 77267-674 77267-676
Allyl phenyl carbonate 97%
Allyl phenyl carbonate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.1846 g/mol
MDL Number: MFCD07784342
CAS Number: 16308-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR