5-Acetyl-2-fluorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (5-Acetyl-2-fluorophenyl)boronic acid

A717861-1G A717861-5G
77280-936EA 19 USD
77280-936 77280-938
5-Acetyl-2-fluorophenylboronic acid 95%
5-Acetyl-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₃
MDL Number: MFCD06656263
CAS Number: 870777-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR