(S)-2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: JQ-1 (carboxylic acid)

A717701-1MG A717701-5MG A717701-100MG A717701-1G A717701-250MG A717701-5G
77243-620EA 6.18 USD
77243-620 77243-622 77552-200 77552-202 77552-204 77552-206
(S)-2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetic acid 98%
(S)-2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇ClN₄O₂S
MW: 400.88 g/mol
MDL Number: MFCD28167916
CAS Number: 202592-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR